Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079420.2 | NP_524144.1 | 2 - 3 | 290 | 93 | |
| GenBank mRNA | AK311942.1 | - | 2 - 3 | 311 | 93 | |
| CR456869.1 | - | 2 - 3 | 93 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002732.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043869.hg.1 | TC02002732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043869.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043869.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | TC02002732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | ENST00000341685.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | ENST00000496436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043868.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043874.hg.1 | TC02002732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043874.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043874.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214477.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214477.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214477.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214477.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214477.hg.1 | TC0200015607.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214478.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214478.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214478.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214478.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214478.hg.1 | TC0200015607.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214471.hg.1 | TC0200015607.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214470.hg.1 | TC0200015607.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |

MYL1
Genomic Map