Assay Details
Gene Symbol: SUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011528575.1 | XP_011526877.1 | 1 - 2 | 91 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528575.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
