Assay Details
| Gene Symbol: SOCS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080867.2 | NP_543143.1 | 2 | 769 | 150 | |
| NM_199421.1 | NP_955453.1 | 3 | 898 | 150 | ||
| XM_011536425.1 | XP_011534727.1 | 3 | 952 | 150 | ||
| XM_011536426.1 | XP_011534728.1 | 3 | 867 | 150 | ||
| GenBank mRNA | AF424815.1 | - | 1 | 549 | 150 | |
| AK124083.1 | - | 2 | 622 | 150 | ||
| BC060790.1 | - | 3 | 776 | 30338646 | 150 | |
| BX161418.1 | - | 2 | 665 | 150 | ||
| BX648734.1 | - | 2 | 555 | 150 | ||
| CR933659.1 | - | 2 | 659 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000321.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of epidermal growth factor-activated receptor activity
protein ubiquitination
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
protein binding
negative regulation of epidermal growth factor-activated receptor activity
protein ubiquitination
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003416.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003419.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003419.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003419.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003419.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003413.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003413.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003413.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003423.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003415.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003415.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003415.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003415.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003415.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003418.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003418.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003418.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003421.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003422.hg.1 | TC14000321.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150044.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150046.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150046.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150046.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150046.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150046.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150041.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150041.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150041.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150041.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150041.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150051.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150050.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007221.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000339298.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150049.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | ENST00000553735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150047.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | ENST00000395472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | ENST00000555846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | NM_080867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | NM_199421.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150043.hg.1 | TC1400007221.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 108883 | - | - |

SOCS4
Genomic Map