Assay Details
Gene Symbol: HIST1H2AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003513.2 | NP_003504.2 | 1 | 433 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812685 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15793601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812679 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 97118 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 17980 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P223384 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P223384 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 44522 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P127214 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P127214 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60812 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P31700934 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P223384 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00812669 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P223384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624454 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624456 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624456 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624454 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624452 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P223384 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P127214 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624443 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624448 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624453 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624458 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624452 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624456 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624460 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624447 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624441 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624450 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624444 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624439 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624437 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04624438 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04624439 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P127214 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P127214 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P223384 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 2142 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 5482 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 8651 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 24767 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P127214 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624384 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624399 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624391 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624447 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793600 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624443 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624450 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624441 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624437 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624458 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624401 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624460 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624439 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P127214 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003513_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124393 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124392 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8124391 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208569_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11741215_at | - | - |
Affymetrix Human Genome U219 Array | 11741215_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208569_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214469_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214469_at | - | - |
Human Genome U133 Plus 2.0 Array | 208569_at | - | - |
Human Genome U133A 2.0 Array | 214469_at | - | - |
Human Genome U133A 2.0 Array | 208569_at | - | - |
Human Genome U133A 2.0 Array | 214469_PM_at | - | - |
Human Genome U133A 2.0 Array | 208569_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003513_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw574298 | - | - |
Human Genome U95 Set | 35127_PM_at | - | - |
Human Genome U95 Set | 35127_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208569_at8 | - | - |
Human Genome U133 Set | 214469_PM_at | - | - |
Human Genome U133 Set | 214469_at | - | - |
Human Genome U133 Set | 208569_PM_at | - | - |
Human Genome U133 Set | 208569_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029525 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 162382 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1753524 | AK311785.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_19557655-S | NM_003513.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_16144-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005130670 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005130670 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_16144 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_16144 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1753524 | BC125140.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1753524 | NM_003513.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1753524 | AK311785.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1753524 | BC125140.1 | human_transcriptome |
