Assay Details
Gene Symbol: CASKIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020764.3 | NP_065815.1 | 11 - 12 | 1184 | 107 | |
GenBank mRNA | AB037727.1 | - | 3 - 4 | 321 | 107 | |
AF451977.1 | - | 11 - 12 | 1250 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000777.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16011428.hg.1 | ENST00000343516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16011428.hg.1 | ENST00000382453.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16011428.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16011428.hg.1 | TC16000777.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600169181.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169181.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169181.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169181.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169181.hg.1 | TC1600009096.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
