| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014083.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014083.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002080.hg.1 |
TC06000223.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002080.hg.1 |
ENST00000359193.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002080.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197767.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197767.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197767.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197767.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197767.hg.1 |
TC0600014083.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014083.hg.1 |
ENST00000359193.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014083.hg.1 |
NM_021064.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014083.hg.1 |
NM_021064.4 |
Refseq |