Assay Details
Gene Symbol: RTN4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023004.5 | NP_075380.1 | 1 - 2 | 228 | 127 | |
GenBank mRNA | AL834449.1 | - | 1 - 2 | 171 | 127 | |
BC011787.1 | - | 1 - 2 | 73 | 4040540 | 127 | |
BM762412.1 | - | 1 - 2 | 228 | 127 | ||
CR456360.1 | - | 1 - 2 | 67 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000508.hg.1
Additional Information:
This assay is in the 5'UTR region of the AL834449.1,BC011787.1,CR456360.1,NM_023004.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
axonogenesis
cytokine-mediated signaling pathway
neuronal signal transduction
negative regulation of axon extension
positive regulation of GTPase activity
negative regulation of JAK-STAT cascade
negative regulation of axonogenesis
protein kinase inhibitor activity
receptor activity
protein binding
neuregulin receptor activity
negative regulation of protein kinase activity
axonogenesis
cytokine-mediated signaling pathway
neuronal signal transduction
negative regulation of axon extension
positive regulation of GTPase activity
negative regulation of JAK-STAT cascade
negative regulation of axonogenesis
protein kinase inhibitor activity
receptor activity
protein binding
neuregulin receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22009410.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009410.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009410.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009410.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009412.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009412.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009412.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009412.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009417.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009417.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009417.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009418.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009418.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009418.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009415.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009415.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009415.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009409.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009409.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009409.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22009411.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009411.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22009411.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22009411.hg.1 | TC22000508.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | TC2200009314.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169709.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009314.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | TC2200009314.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000463936.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169711.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169715.hg.1 | TC2200009314.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169715.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169715.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169715.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169715.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | TC2200009314.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000416372.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000425986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | ENST00000469601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169710.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169716.hg.1 | TC2200009314.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169716.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169716.hg.1 | NM_023004.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169716.hg.1 | ENST00000043402.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169716.hg.1 | NM_023004.5 | Refseq |
