Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025151.4 | NP_079427.4 | 3 - 4 | 1682 | 117 | |
GenBank mRNA | AF368294.1 | - | 3 - 4 | 1652 | 117 | |
AK092296.1 | - | 2 - 3 | 990 | 117 | ||
AK291781.1 | - | 3 - 4 | 1679 | 117 | ||
AY280970.1 | - | 2 - 3 | 1815 | 117 | ||
BC077720.1 | - | 3 - 4 | 1652 | 6537110 | 117 | |
DQ083995.1 | - | 3 - 4 | 1406 | 117 | ||
DQ236342.1 | - | 3 - 4 | 1652 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001138.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | TC08001138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | ENST00000287263.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | ENST00000330843.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | ENST00000522727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | ENST00000523182.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08014594.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168922.hg.1 | TC0800010138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 220072 | - | - |
