Assay Details
Gene Symbol: LRRC46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033413.3 | NP_219481.1 | 1 - 2 | 366 | 98 | |
GenBank mRNA | AK027206.1 | - | 1 - 2 | 270 | 99 | |
AK292765.1 | - | 1 - 2 | 281 | 98 | ||
AK315665.1 | - | 1 - 2 | 366 | 98 | ||
BC010412.2 | - | 1 - 2 | 259 | 3836250 | 98 | |
DB024018.1 | - | 1 - 2 | 366 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000614.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292765.1,AK315665.1,BC010412.2,NM_033413.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17007589.hg.1 | ENST00000269025.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007589.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007589.hg.1 | TC17000614.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17007590.hg.1 | ENST00000269025.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007590.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007590.hg.1 | TC17000614.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007591.hg.1 | ENST00000269025.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007591.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007591.hg.1 | TC17000614.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159930.hg.1 | TC1700008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159928.hg.1 | TC1700008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159926.hg.1 | TC1700008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159925.hg.1 | TC1700008138.hg.1 | Clariom D TC |
