Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003498.hg.1 |
ENST00000292431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003498.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003498.hg.1 |
TC19000236.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003499.hg.1 |
ENST00000292431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003499.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003499.hg.1 |
TC19000236.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000586171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
ENST00000586171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151281.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151281.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151281.hg.1 |
TC1900007125.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900151281.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151281.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
TC1900007125.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151280.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000292431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000585663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000586171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
ENST00000586171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007125.hg.1 |
NM_052876.3 |
Refseq |