Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256409.1 | NP_001243338.1 | 4 - 5 | 879 | 104 | |
NM_052940.4 | NP_443172.1 | 3 - 4 | 845 | 104 | ||
XM_006710327.3 | XP_006710390.1 | 4 - 5 | 701 | 104 | ||
XM_006710328.3 | XP_006710391.1 | 4 - 5 | 1214 | 104 | ||
GenBank mRNA | AK074521.1 | - | 3 - 4 | 818 | 104 | |
AK075201.1 | - | 3 - 4 | 807 | 104 | ||
AK090881.1 | - | 4 - 5 | 1128 | 104 | ||
AK095767.1 | - | 4 - 5 | 798 | 104 | ||
BC013101.1 | - | 3 - 4 | 684 | 3918785 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000659.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01010710.hg.1 | TC01000659.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010710.hg.1 | ENST00000319223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010710.hg.1 | ENST00000371370.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010710.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010710.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010709.hg.1 | TC01000659.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010709.hg.1 | ENST00000319223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010709.hg.1 | ENST00000371370.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010709.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010709.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159608.hg.1 | TC0100008369.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159609.hg.1 | TC0100008369.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
