Assay Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080476.4 | NP_536724.1 | 10 - 11 | 1251 | 61 | |
| XM_011528542.2 | XP_011526844.1 | 4 - 5 | 542 | 61 | ||
| XM_017027664.1 | XP_016883153.1 | 9 - 10 | 925 | 61 | ||
| GenBank mRNA | AB086842.1 | - | 10 - 11 | 1057 | 61 | |
| AK075507.1 | - | 10 - 11 | 1076 | 61 | ||
| AK301900.1 | - | 10 - 11 | 1051 | 61 | ||
| AY339061.1 | - | 9 - 10 | 991 | 61 | ||
| AY358816.1 | - | 10 - 11 | 1081 | 61 | ||
| AY422169.1 | - | 10 - 11 | 1251 | 61 | ||
| BC030512.1 | - | 10 - 11 | 1062 | 4942673 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000784.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20010451.hg.1 | ENST00000217446.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010451.hg.1 | ENST00000374820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010451.hg.1 | ENST00000452740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010451.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010451.hg.1 | TC20000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | TC2000010001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171368.hg.1 | NM_080476.4 | Refseq |

PIGU
Genomic Map