Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167076.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167076.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167076.hg.1 |
TC2000009618.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000167076.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167076.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167075.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167075.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167075.hg.1 |
TC2000009618.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000167075.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167075.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
ENST00000457363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009618.hg.1 |
NM_001304369.1 |
Refseq |