Assay Details
Gene Symbol: GPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142466.2 | NP_001135938.1 | 4 - 5 | 528 | 63 | |
NM_133443.3 | NP_597700.1 | 4 - 5 | 552 | 63 | ||
GenBank mRNA | AK094971.1 | - | 4 - 5 | 452 | 63 | |
AY029173.2 | - | 4 - 5 | 536 | 63 | ||
BC062555.1 | - | 4 - 5 | 501 | 6429257 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000426.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16005890.hg.1 | ENST00000340124.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16005890.hg.1 | ENST00000440783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16005890.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16005890.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16005890.hg.1 | TC16000426.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156353.hg.1 | TC1600007723.hg.1 | Clariom D TC |
