| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
TC06000737.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
ENST00000287097.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
ENST00000422508.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
ENST00000437994.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
NM_001159587.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
NM_001159588.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008876.hg.1 |
NM_133493.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
TC06000737.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
ENST00000287097.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
ENST00000422508.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
ENST00000437994.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
ENST00000474094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
NM_001159587.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
NM_001159588.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008877.hg.1 |
NM_133493.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000287097.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000422508.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000437994.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_001159587.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_001159588.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_133493.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000287097.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000422508.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
ENST00000437994.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_001159587.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_001159588.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
NM_133493.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600159407.hg.1 |
TC0600008539.hg.1 |
Clariom D TC |