Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138410.3 | NP_612419.1 | 2 - 3 | 570 | 67 | |
XM_011533319.2 | XP_011531621.1 | 2 - 3 | 396 | 67 | ||
XM_017005646.1 | XP_016861135.1 | 2 - 3 | 416 | 67 | ||
GenBank mRNA | AF479263.2 | - | 2 - 3 | 570 | 67 | |
BC010116.2 | - | 2 - 3 | 352 | 3636045 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000161.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03002828.hg.1 | TC03000161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03002828.hg.1 | ENST00000334983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03002828.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148470.hg.1 | TC0300006968.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
