Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138410.3 | NP_612419.1 | 3 - 4 | 669 | 71 | |
GenBank mRNA | AF479263.2 | - | 3 - 4 | 669 | 71 | |
BC010116.2 | - | 3 - 4 | 451 | 3636045 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000161.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03002829.hg.1 | TC03000161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03002829.hg.1 | ENST00000334983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03002829.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03002833.hg.1 | TC03000161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03002833.hg.1 | ENST00000334983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03002833.hg.1 | ENST00000349718.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03002833.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03002833.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000465248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006968.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000349718.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000464689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | NM_181472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148475.hg.1 | TC0300006968.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000334983.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000454304.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | ENST00000487007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | NM_138410.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148471.hg.1 | TC0300006968.hg.1 | Clariom D TC |
