Assay Details
| Gene Symbol: PPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL110128.1 | - | 1 - 2 | 114 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AL110128.1 transcripts.
Set Membership: |
|

PPT2
Genomic Map