Assay Details
Gene Symbol: HIST1H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003522.3 | NP_003513.1 | 1 | 356 | 84 | |
GenBank mRNA | BC096120.1 | - | 1 | 356 | 40002352 | 84 |
BC096123.1 | - | 1 | 356 | 40002357 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000171.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15793718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813477 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3229122 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P42178 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 16616 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P101509 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P101509 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15040 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P37554978 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3229122 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3229122 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15793718 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P42178 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P101509 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15793722 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P101509 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P42178 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 7315 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 97092 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P101509 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793716 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625226 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793717 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625241 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625252 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625250 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625232 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625243 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793734 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625237 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625246 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625228 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625239 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625234 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003522_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117401 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117397 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117396 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117398 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117399 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117400 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117395 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208490_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11715118_s_at | - | - |
Affymetrix Human Genome U219 Array | 11759156_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11759157_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11759156_at | - | - |
Affymetrix Human Genome U219 Array | 11715118_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11759157_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06001746.hg.1 | TC06000171.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001746.hg.1 | ENST00000359985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001746.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001747.hg.1 | TC06000171.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001747.hg.1 | ENST00000359985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001747.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007282.hg.1 | ENST00000356530.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007282.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007282.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149353.hg.1 | ENST00000356530.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149353.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149353.hg.1 | ENST00000356530.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149353.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149353.hg.1 | TC0600007282.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149354.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149354.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149354.hg.1 | TC0600007282.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007282.hg.1 | ENST00000356530.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007282.hg.1 | NM_003522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007282.hg.1 | NM_003522.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 215779_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 208523_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 210387_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208490_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208523_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 210387_at | - | - |
Human Genome U133 Plus 2.0 Array | 214455_at | - | - |
Human Genome U133 Plus 2.0 Array | 208527_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208490_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 215779_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 214455_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208527_x_at | - | - |
Human Genome U133A 2.0 Array | 215779_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 208490_x_at | - | - |
Human Genome U133A 2.0 Array | 214455_at | - | - |
Human Genome U133A 2.0 Array | 208490_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208527_x_at | - | - |
Human Genome U133A 2.0 Array | 215779_s_at | - | - |
Human Genome U133A 2.0 Array | 208523_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 210387_at | - | - |
Human Genome U133A 2.0 Array | 208527_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 210387_PM_at | - | - |
Human Genome U133A 2.0 Array | 208523_x_at | - | - |
Human Genome U133A 2.0 Array | 214455_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003522_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw563155 | - | - |
Human Genome U95 Set | 32980_PM_f_at | - | - |
Human Genome U95 Set | 31524_PM_f_at | - | - |
Human Genome U95 Set | 31522_PM_f_at | - | - |
Human Genome U95 Set | 31523_PM_f_at | - | - |
Human Genome U95 Set | 32980_f_at | - | - |
Human Genome U95 Set | 31523_f_at | - | - |
Human Genome U95 Set | 31522_f_at | - | - |
Human Genome U95 Set | 31524_f_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208490_x_at11 | - | - |
Human Genome U95 Set | 55167_r_at | - | - |
Human Genome U95 Set | 55167_PM_r_at | - | - |
Human Genome U133 Set | 215779_PM_s_at | - | - |
Human Genome U133 Set | 208490_x_at | - | - |
Human Genome U133 Set | 208490_PM_x_at | - | - |
Human Genome U133 Set | 210387_at | - | - |
Human Genome U133 Set | 208527_x_at | - | - |
Human Genome U133 Set | 210387_PM_at | - | - |
Human Genome U133 Set | 214455_at | - | - |
Human Genome U133 Set | 215779_s_at | - | - |
Human Genome U133 Set | 214455_PM_at | - | - |
Human Genome U133 Set | 208523_x_at | - | - |
Human Genome U133 Set | 208523_PM_x_at | - | - |
Human Genome U133 Set | 208527_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031108 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031108 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 103461 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1779373 | BC056264.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1779373 | BC096120.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21396482-S | NM_003522.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21396482-S | BC056264.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21396482-S | BC096120.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21396482-S | BC096123.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27586-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001050427 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001050427 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1780047 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27586 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1779373 | BC096123.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1779373 | NM_003522.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1779373 | BC056264.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1779373 | BC096120.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1779373 | BC096123.1 | human_transcriptome |
