Assay Details
| Gene Symbol: DCHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003737.3 | NP_003728.1 | 2 - 3 | 2205 | 56 | |
| GenBank mRNA | AB053446.1 | - | 2 - 3 | 2208 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001363.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
mitral valve formation
cell migration involved in endocardial cushion formation
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
neural tube development
neurogenesis
hippo signaling
post-anal tail morphogenesis
ossification involved in bone maturation
digestive tract development
cochlea development
calcium ion binding
branching involved in ureteric bud morphogenesis
mitral valve formation
cell migration involved in endocardial cushion formation
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
neural tube development
neurogenesis
hippo signaling
post-anal tail morphogenesis
ossification involved in bone maturation
digestive tract development
cochlea development
calcium ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11017185.hg.1 | ENST00000299441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017185.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017185.hg.1 | TC11001363.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017183.hg.1 | ENST00000299441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017183.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017183.hg.1 | TC11001363.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017184.hg.1 | ENST00000299441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017184.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017184.hg.1 | TC11001363.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100175472.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175472.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175472.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175472.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175472.hg.1 | TC1100009982.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100175474.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175474.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175474.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175474.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175474.hg.1 | TC1100009982.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009982.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175473.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175473.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175473.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175473.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175473.hg.1 | TC1100009982.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100175471.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175471.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175471.hg.1 | ENST00000299441.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175471.hg.1 | NM_003737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175471.hg.1 | TC1100009982.hg.1 | Clariom D TC |

DCHS1
Genomic Map