Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014878.4 | NP_055693.4 | 7 - 8 | 771 | 136 | |
GenBank mRNA | AF490254.1 | - | 4 - 5 | 255 | 136 | |
AK225953.1 | - | 7 - 8 | 736 | 136 | ||
AK292169.1 | - | 7 - 8 | 714 | 136 | ||
AK315531.1 | - | 7 - 8 | 750 | 136 | ||
AL832239.1 | - | 7 - 8 | 737 | 136 | ||
BC016137.2 | - | 6 - 7 | 675 | 3908516 | 136 | |
CR456957.1 | - | 4 - 5 | 255 | 136 | ||
D13645.1 | - | 6 - 7 | 673 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000874.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09011627.hg.1 | TC09000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011627.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011627.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09011628.hg.1 | TC09000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011628.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011628.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09011631.hg.1 | TC09000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011631.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011631.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164598.hg.1 | TC0900009404.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164597.hg.1 | TC0900009404.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900164600.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164600.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164600.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900164600.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900164600.hg.1 | TC0900009404.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000382032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000397885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000469168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | ENST00000490444.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009404.hg.1 | NM_014878.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007163.hg.1 | ENST00000565135.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175386 | - | - |
