Assay Details
Gene Symbol: PPP4R4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058237.1 | NP_478144.1 | 10 - 11 | 1300 | 153 | |
XM_005267930.2 | XP_005267987.1 | 10 - 11 | 1133 | 153 | ||
XM_011537038.2 | XP_011535340.1 | 10 - 11 | 1296 | 153 | ||
XM_011537039.2 | XP_011535341.1 | 9 - 10 | 1041 | 153 | ||
XM_011537040.2 | XP_011535342.1 | 8 - 9 | 948 | 153 | ||
XM_017021528.1 | XP_016877017.1 | 8 - 9 | 985 | 153 | ||
XM_017021529.1 | XP_016877018.1 | 12 - 13 | 1655 | 153 | ||
XM_017021530.1 | XP_016877019.1 | 12 - 13 | 1629 | 153 | ||
GenBank mRNA | AB046842.1 | - | 10 - 11 | 1219 | 153 | |
BC068491.1 | - | 10 - 11 | 1259 | 5297335 | 153 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000585.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14007230.hg.1 | ENST00000304338.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14007230.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14007230.hg.1 | TC14000585.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400157181.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400157181.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400157181.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400157181.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400157181.hg.1 | TC1400008058.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
