Assay Details
Gene Symbol: RXFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166058.1 | NP_001159530.1 | 7 - 8 | 706 | 88 | |
NM_130806.3 | NP_570718.1 | 7 - 8 | 706 | 88 | ||
XM_017020389.1 | XP_016875878.1 | 7 - 8 | 695 | 88 | ||
GenBank mRNA | AF403384.2 | - | 7 - 8 | 741 | 88 | |
AF453828.1 | - | 7 - 8 | 705 | 88 | ||
AY899851.1 | - | 7 - 8 | 635 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000106.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oocyte maturation
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
male gonad development
positive regulation of cAMP biosynthetic process
negative regulation of apoptotic process
G-protein coupled receptor activity
protein-hormone receptor activity
peptide hormone binding
oocyte maturation
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
male gonad development
positive regulation of cAMP biosynthetic process
negative regulation of apoptotic process
G-protein coupled receptor activity
protein-hormone receptor activity
peptide hormone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001037.hg.1 | ENST00000298386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001037.hg.1 | ENST00000380314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001037.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001037.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001037.hg.1 | TC13000106.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006794.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | ENST00000298386.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | ENST00000380314.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | NM_001166058.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | NM_130806.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146149.hg.1 | TC1300006794.hg.1 | Clariom D TC |
