Assay Details
| Gene Symbol: CRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139161.4 | NP_631900.1 | 1 - 2 | 113 | 90 | |
| NM_174881.3 | NP_777377.1 | 1 - 2 | 113 | 90 | ||
| GenBank mRNA | AF503290.1 | - | 1 - 2 | 72 | 90 | |
| AK000653.1 | - | 1 - 2 | 52 | 90 | ||
| AK292956.1 | - | 1 - 2 | 78 | 90 | ||
| AY358684.1 | - | 1 - 2 | 68 | 90 | ||
| BC018409.1 | - | 1 - 2 | 64 | 3861124 | 90 | |
| BM129809.1 | - | 1 - 2 | 81 | 90 | ||
| CN403242.1 | - | 1 - 2 | 102 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000115.hg
Additional Information:
This assay is in the 5'UTR region of the AF503290.1,AK292956.1,AY358684.1,BC018409.1,NM_139161.4,NM_174881.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19001741.hg.1 | ENST00000308243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001741.hg.1 | ENST00000356762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001741.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001741.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001741.hg.1 | TC19000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001739.hg.1 | ENST00000308243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001739.hg.1 | ENST00000356762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001739.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001739.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001739.hg.1 | TC19000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001737.hg.1 | ENST00000356762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001737.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001737.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001737.hg.1 | TC19000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001740.hg.1 | ENST00000308243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001740.hg.1 | ENST00000356762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001740.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001740.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001740.hg.1 | TC19000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | TC1900006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147768.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006801.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | TC1900006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147766.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | TC1900006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000308243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000598494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147767.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147761.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147761.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147761.hg.1 | TC1900006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147761.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147761.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | NM_174881.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | TC1900006801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | ENST00000356762.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | ENST00000600229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | NM_139161.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147764.hg.1 | NM_174881.3 | Refseq |

CRB3
Genomic Map