Assay Details
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139278.2 | NP_644807.1 | 6 - 7 | 953 | 78 | |
GenBank mRNA | AF467956.1 | - | 6 - 7 | 874 | 78 | |
AJ487518.1 | - | 6 - 7 | 1083 | 78 | ||
AJ487961.1 | - | 6 - 7 | 953 | 78 | ||
AK090946.1 | - | 3 - 4 | 274 | 78 | ||
AY358116.1 | - | 6 - 7 | 691 | 78 | ||
BC142997.1 | - | 5 - 6 | 801 | 8860454 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001035.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK090946.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013210.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013210.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013210.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013210.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013209.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013209.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013209.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013209.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166783.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166782.hg.1 | TC0800009831.hg.1 | Clariom D TC |
