Assay Details
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002135.4 | NP_002126.2 | 2 - 3 | 317 | 96 | |
XM_006719363.1 | XP_006719426.1 | 2 - 3 | 223 | 96 | ||
GenBank mRNA | BC016147.1 | - | 2 - 3 | 262 | 3921259 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC016147.1, NM_002135.4, XM_006719363.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
