Assay Details
Gene Symbol: DHX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003587.4 | NP_003578.2 | 1 - 2 | 417 | 72 | |
GenBank mRNA | AB001601.1 | - | 1 - 2 | 344 | 72 | |
AK312264.1 | - | 1 - 2 | 356 | 72 | ||
BC002789.2 | - | 1 - 2 | 340 | 72 | ||
BC008825.2 | - | 1 - 2 | 368 | 4100320 | 72 | |
BC009392.2 | - | 1 - 2 | 368 | 4136069 | 72 | |
DB072495.1 | - | 1 - 2 | 418 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001504.hg.1, TC6_apd_hap1000074.hg.1, TC6_cox_hap2000138.hg.1, TC6_dbb_hap3000127.hg.1, TC6_mann_hap4000116.hg.1, TC6_mcf_hap5000117.hg.1, TC6_qbl_hap6000128.hg.1, TC6_ssto_hap7000119.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06016995.hg.1 | TC06001504.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016995.hg.1 | ENST00000376442.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016995.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016995.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002119.hg.1 | TC6_mcf_hap5000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002119.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002119.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002403.hg.1 | TC6_qbl_hap6000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002403.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001950.hg.1 | TC6_mann_hap4000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001950.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002123.hg.1 | TC6_mcf_hap5000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002123.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002123.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002402.hg.1 | TC6_qbl_hap6000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002402.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002402.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002204.hg.1 | TC6_ssto_hap7000119.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002204.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001028.hg.1 | TC6_apd_hap1000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001028.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002430.hg.1 | TC6_cox_hap2000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002430.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002359.hg.1 | TC6_dbb_hap3000127.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002359.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002359.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002120.hg.1 | TC6_mcf_hap5000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002120.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002203.hg.1 | TC6_ssto_hap7000119.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002203.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002203.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016993.hg.1 | TC06001504.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016993.hg.1 | ENST00000376442.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016993.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016993.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001031.hg.1 | TC6_apd_hap1000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001031.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001031.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002429.hg.1 | TC6_cox_hap2000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002429.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002429.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002121.hg.1 | TC6_mcf_hap5000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002121.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002121.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016991.hg.1 | TC06001504.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016991.hg.1 | ENST00000376442.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016991.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016991.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001029.hg.1 | TC6_apd_hap1000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001029.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001029.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001949.hg.1 | TC6_mann_hap4000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001949.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001949.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016992.hg.1 | TC06001504.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016992.hg.1 | ENST00000376442.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016992.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001027.hg.1 | TC6_apd_hap1000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001027.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001027.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002360.hg.1 | TC6_dbb_hap3000127.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002360.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176684.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176684.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176684.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176684.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176684.hg.1 | TC0600011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600176687.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176687.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176687.hg.1 | TC0600011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600176686.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176686.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176686.hg.1 | TC0600011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 120967 | - | - |
