Assay Details
Gene Symbol: CHST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024533.4 | NP_078809.2 | 2 - 3 | 138 | 70 | |
GenBank mRNA | AF246718.1 | - | 2 - 3 | 138 | 70 | |
BX282540.1 | - | 2 - 3 | 215 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16002090.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF246718.1,NM_024533.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017838.hg.1 | TC16002090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017838.hg.1 | ENST00000336257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017838.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017836.hg.1 | TC16002090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017836.hg.1 | ENST00000336257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017836.hg.1 | ENST00000541075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017836.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017837.hg.1 | TC16002090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017837.hg.1 | ENST00000336257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017837.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017839.hg.1 | TC16002090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017839.hg.1 | ENST00000336257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017839.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017840.hg.1 | TC16002090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017840.hg.1 | ENST00000336257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017840.hg.1 | ENST00000541075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017840.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194487.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194487.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194487.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194487.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194487.hg.1 | TC1600011567.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600194489.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194489.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194489.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194489.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194489.hg.1 | TC1600011567.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194486.hg.1 | TC1600011567.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600194488.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194488.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194488.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194488.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194488.hg.1 | TC1600011567.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000332272.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | ENST00000390664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010881.hg.1 | NM_021615.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | ENST00000336257.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | ENST00000565039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | NM_024533.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194490.hg.1 | TC1600011567.hg.1 | Clariom D TC |
