Assay Details
Gene Symbol: MORC4 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085354.2 | NP_001078823.1 | 7 - 8 | 1211 | 82 | ||
NM_024657.4 | NP_078933.3 | 7 - 8 | 1211 | 82 | |||
XM_005262190.3 | XP_005262247.1 | 6 - 7 | 1101 | 82 | |||
XM_006724691.2 | XP_006724754.1 | 5 - 6 | 845 | 82 | |||
XM_011531027.2 | XP_011529329.1 | 4 - 5 | 499 | 82 | |||
XM_017029844.1 | XP_016885333.1 | 6 - 7 | 1101 | 82 | |||
GenBank mRNA | AK056235.1 | - | 3 - 4 | 423 | 82 | ||
AK300305.1 | - | 6 - 7 | 786 | 82 | |||
AY927537.1 | - | LIT3543 AY927537 |
5 - 6 | 746 | 82 | ||
EF125209.1 | - | 7 - 8 | 936 | 82 | |||
EF125210.1 | - | 6 - 7 | 995 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001257.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | TC0X001257.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | ENST00000355610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | ENST00000535534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016967.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171227.hg.1 | TC0X00010459.hg.1 | Clariom D TC |
