Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303126.1 | NP_001290055.1 | 5 - 6 | 643 | 86 | |
| NM_144593.2 | NP_653194.1 | 5 - 6 | 575 | 86 | ||
| NR_130123.1 | 4 - 5 | 507 | 86 | |||
| XM_017018794.1 | XP_016874283.1 | 5 - 6 | 1232 | 86 | ||
| XM_017018795.1 | XP_016874284.1 | 5 - 6 | 1300 | 86 | ||
| GenBank mRNA | AK098663.1 | - | 5 - 6 | 574 | 86 | |
| AY327412.1 | - | 5 - 6 | 575 | 86 | ||
| AY509932.1 | - | 5 - 6 | 641 | 86 | ||
| BC014155.2 | - | 4 - 5 | 429 | 86 | ||
| BC027482.1 | - | 5 - 6 | 542 | 4396827 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001467.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12019274.hg.1 | ENST00000301068.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019274.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019274.hg.1 | TC12001467.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180586.hg.1 | TC1200010612.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180587.hg.1 | TC1200010612.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000301068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000420065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | ENST00000550797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_001303126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NM_144593.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010612.hg.1 | NR_130123.1 | Refseq |

RHEBL1
Genomic Map