Assay Details
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144647.3 | NP_653248.3 | 1 - 2 | 126 | 99 | |
| GenBank mRNA | BC017586.2 | - | 1 - 2 | 128 | 4837506 | 99 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001265.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC017586.2,NM_144647.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017690.hg.1 | TC05001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017690.hg.1 | ENST00000397367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017690.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | TC05001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | ENST00000397366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | ENST00000397367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | ENST00000514524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017688.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172979.hg.1 | TC0500010489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500172981.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172981.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172981.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172981.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172981.hg.1 | TC0500010489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000397367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000513623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | ENST00000514524.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_001042625.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010489.hg.1 | NM_144647.3 | Refseq |

CAPSL
Genomic Map