Assay Details
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144673.2 | NP_653274.1 | 1 - 2 | 443 | 65 | |
GenBank mRNA | AF479260.1 | - | 1 - 2 | 435 | 65 | |
AK098598.1 | - | 1 - 2 | 428 | 65 | ||
BC025354.1 | - | 1 - 2 | 434 | 4904062 | 65 | |
BI830241.1 | - | 1 - 2 | 443 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000519.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16007246.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16007246.hg.1 | ENST00000379486.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16007246.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16007246.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16007246.hg.1 | TC16000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16007247.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16007247.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16007247.hg.1 | TC16000519.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600159888.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159888.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159888.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159888.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159888.hg.1 | TC1600008126.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159886.hg.1 | TC1600008126.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159885.hg.1 | TC1600008126.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000532362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | ENST00000569316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008126.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | ENST00000268595.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | ENST00000379486.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | NM_001199317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | NM_144673.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600159887.hg.1 | TC1600008126.hg.1 | Clariom D TC |
