Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
ENST00000310715.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
ENST00000340329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
ENST00000355577.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
ENST00000394865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
ENST00000401621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
NM_145170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017688.hg.1 |
TC10001406.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000310715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000340329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000355577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000394865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000433268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000462684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000495161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
NM_145170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000310715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000340329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000355577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000394865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000433268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000462684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
ENST00000495161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
NM_145170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174966.hg.1 |
TC1000011017.hg.1 |
Clariom D TC |