Assay Details
Gene Symbol: DNAJB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145174.1 | NP_660157.1 | 1 | 348 | 120 | |
GenBank mRNA | AF085232.1 | - | 1 | 348 | 120 | |
BC112135.1 | - | 1 | 307 | 8327603 | 120 | |
CR456468.1 | - | 1 | 253 | 120 |
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204827.1 | NP_001191756.1 | 3 | 120 | ||
GenBank mRNA | BC005331.1 | - | 3 | 120 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000327.hg.1, TC22000817.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22013877.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22013877.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22013877.hg.1 | TC22000817.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22013878.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22013878.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22013878.hg.1 | TC22000817.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006304.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006304.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006304.hg.1 | TC22000327.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22006303.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006303.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006303.hg.1 | TC22000327.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200169074.hg.1 | TC2200009277.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169074.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169074.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169074.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169074.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008800.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163782.hg.1 | TC2200008800.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200163782.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163782.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163782.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163782.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163781.hg.1 | TC2200008800.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200163781.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163781.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163781.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163781.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163783.hg.1 | TC2200008800.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200163783.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163783.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163783.hg.1 | ENST00000307221.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163783.hg.1 | NM_145174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | TC2200009277.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | NM_001204827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | ENST00000482652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | ENST00000614001.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200169072.hg.1 | NM_001204827.1 | Refseq |
