Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016731.hg.1 |
ENST00000293725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016731.hg.1 |
ENST00000318168.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016731.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016731.hg.1 |
TC19001196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016732.hg.1 |
ENST00000318168.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016732.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016732.hg.1 |
TC19001196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016730.hg.1 |
ENST00000293725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016730.hg.1 |
ENST00000318168.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016730.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016730.hg.1 |
TC19001196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016728.hg.1 |
ENST00000293725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016728.hg.1 |
ENST00000318168.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016728.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016728.hg.1 |
TC19001196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016729.hg.1 |
ENST00000293725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016729.hg.1 |
ENST00000318168.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016729.hg.1 |
TC19001196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
TC1900009729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179074.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
TC1900009729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179072.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009729.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
TC1900009729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179079.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
TC1900009729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000293725.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000594577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
ENST00000601858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179073.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179080.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179080.hg.1 |
NM_145276.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179080.hg.1 |
TC1900009729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179080.hg.1 |
ENST00000595977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179080.hg.1 |
NM_145276.2 |
Refseq |