Assay Details
Gene Symbol: MAP4K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242559.1 | NP_001229488.1 | 16 - 17 | 1867 | 81 | |
NM_145686.3 | NP_663719.2 | 16 - 17 | 2147 | 81 | ||
NM_145687.3 | NP_663720.1 | 16 - 17 | 1774 | 81 | ||
XM_005264057.3 | XP_005264114.1 | 16 - 17 | 1936 | 81 | ||
XM_005264058.3 | XP_005264115.1 | 16 - 17 | 1936 | 81 | ||
XM_005264059.3 | XP_005264116.1 | 16 - 17 | 1843 | 81 | ||
XM_005264060.3 | XP_005264117.1 | 16 - 17 | 1843 | 81 | ||
XM_005264069.2 | XP_005264126.1 | 16 - 17 | 1843 | 81 | ||
XM_017005351.1 | XP_016860840.1 | 16 - 17 | 1936 | 81 | ||
XM_017005352.1 | XP_016860841.1 | 16 - 17 | 1936 | 81 | ||
XM_017005353.1 | XP_016860842.1 | 16 - 17 | 1936 | 81 | ||
XM_017005356.1 | XP_016860845.1 | 16 - 17 | 1843 | 81 | ||
XM_017005357.1 | XP_016860846.1 | 16 - 17 | 1843 | 81 | ||
XM_017005358.1 | XP_016860847.1 | 16 - 17 | 1843 | 81 | ||
XM_017005365.1 | XP_016860854.1 | 16 - 17 | 1936 | 81 | ||
XM_017005368.1 | XP_016860857.1 | 16 - 17 | 1843 | 81 | ||
GenBank mRNA | AB014587.1 | - | 14 - 15 | 1658 | 81 | |
AY212247.1 | - | 16 - 17 | 1862 | 81 | ||
EU794634.1 | - | 16 - 17 | 1824 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
