Assay Details
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321213.1 | NP_001308142.1 | 1 - 2 | 141 | 134 | |
NM_024963.5 | NP_079239.3 | 1 - 2 | 141 | 134 | ||
XM_011515531.2 | XP_011513833.1 | 1 - 2 | 156 | 134 | ||
XM_011515532.2 | XP_011513834.1 | 1 - 2 | 156 | 134 | ||
GenBank mRNA | AK294949.1 | - | 1 - 2 | 135 | 134 | |
BC006426.1 | - | 1 - 2 | 111 | 4026501 | 134 | |
BC024256.1 | - | 1 - 2 | 111 | 3954975 | 134 | |
DC307306.1 | - | 1 - 2 | 141 | 134 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001118.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294949.1,BC006426.1,BC024256.1,NM_001321213.1,NM_024963.5,XM_011515531.2,XM_011515532.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07017227.hg.1 | TC07001118.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07017227.hg.1 | ENST00000312577.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017227.hg.1 | ENST00000382368.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017227.hg.1 | ENST00000453700.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017227.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07017228.hg.1 | TC07001118.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07017228.hg.1 | ENST00000312577.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017228.hg.1 | ENST00000382368.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017228.hg.1 | ENST00000453700.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017228.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172195.hg.1 | TC0700010162.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172197.hg.1 | TC0700010162.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
