Assay Details
| Gene Symbol: FBXL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321213.1 | NP_001308142.1 | 2 - 3 | 360 | 64 | |
| NM_024963.5 | NP_079239.3 | 2 - 3 | 360 | 64 | ||
| XM_011515531.2 | XP_011513833.1 | 2 - 3 | 375 | 64 | ||
| XM_011515532.2 | XP_011513834.1 | 2 - 3 | 375 | 64 | ||
| GenBank mRNA | AK057042.1 | - | 2 - 3 | 64 | ||
| AK294949.1 | - | 2 - 3 | 64 | |||
| BC006426.1 | - | 2 - 3 | 330 | 4026501 | 64 | |
| BC024256.1 | - | 2 - 3 | 330 | 3954975 | 64 | |
| DC307306.1 | - | 2 - 3 | 64 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001118.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07017226.hg.1 | TC07001118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017226.hg.1 | ENST00000312577.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017226.hg.1 | ENST00000382368.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017226.hg.1 | ENST00000453700.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017226.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017225.hg.1 | TC07001118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017225.hg.1 | ENST00000312577.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017225.hg.1 | ENST00000382368.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017225.hg.1 | ENST00000453700.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017225.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172193.hg.1 | TC0700010162.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172194.hg.1 | TC0700010162.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 153602 | - | - |

FBXL18
Genomic Map