Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019367.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019373.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019370.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019368.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019369.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
ENST00000371426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
ENST00000371430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
ENST00000394100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
ENST00000536233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019372.hg.1 |
TC10001538.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000177773.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177773.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177773.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177773.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177773.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177771.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177771.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177771.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177771.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177771.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177772.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177772.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177772.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177772.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177772.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177776.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177776.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177776.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177776.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177776.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177775.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177775.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177775.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177775.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177775.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177774.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177774.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177774.hg.1 |
ENST00000359204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177774.hg.1 |
NM_145246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177774.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |