Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002385.hg.1 |
TC08000184.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002385.hg.1 |
ENST00000313219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002385.hg.1 |
ENST00000397677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002385.hg.1 |
ENST00000520102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002385.hg.1 |
NM_152272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000313219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000397677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000517325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000520102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000521656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000523091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
NM_152272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000313219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000397677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000517325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000520102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000521656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
ENST00000523091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
NM_152272.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148004.hg.1 |
TC0800007013.hg.1 |
Clariom D TC |