Assay Details
| Gene Symbol: RAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024028.1 | XP_016879517.1 | 1 - 2 | 142 | 87 | |
| GenBank mRNA | BC021209.2 | - | 1 - 2 | 87 | 4137399 | 87 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC021209.2, XM_017024028.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
circadian regulation of gene expression
negative regulation of multicellular organism growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
enhancer binding
skeletal system development
circadian regulation of gene expression
negative regulation of multicellular organism growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
enhancer binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700149878.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149878.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149878.hg.1 | TC1700007080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000353383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000395774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000471135.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000489697.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000582514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | ENST00000583166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007080.hg.1 | NM_030665.3 | Refseq |

RAI1
Genomic Map