Assay Details
| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024470.1 | 3 - 4 | 964 | 99 | ||
| GenBank mRNA | BC025345.1 | - | 3 - 4 | 964 | 4862156 | 99 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001015.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381235 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4592 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4591 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38621 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381235 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3381235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261703 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17208900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912316 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17208900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261703 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11261707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31912316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261696 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261693 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261694 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261738 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261616 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261686 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261711 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261618 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261682 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261621 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17208900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261701 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17208897 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261736 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261722 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11261681 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8063932 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8067548 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8063931 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8067547 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8067549 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8067550 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8067546 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1554332_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11757046_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11757046_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013923.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013923.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013923.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013923.hg.1 | TC20001015.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013922.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013922.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013922.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013922.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013922.hg.1 | TC20001015.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | TC2000009768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168122.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | TC2000009768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168123.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000411824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000433126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | ENST00000451648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009768.hg.1 | NR_024470.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017889 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 217299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3243616 | NR_024470.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3243616 | BC025345.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3243616 | BU741282.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243616 | NR_024470.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243616 | BC025345.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243616 | BU741282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243616 | NR_024470.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243616 | BC025345.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243616 | BU741282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243616 | NR_024470.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243616 | BC025345.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243616 | BU741282.1 | human_transcriptome |

SLCO4A1-AS1
Genomic Map