Assay Details
Gene Symbol: C11orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152587.3 | NP_689800.3 | 3 - 4 | 243 | 70 | |
XM_005271414.3 | XP_005271471.1 | 3 - 4 | 259 | 70 | ||
XM_005271415.4 | XP_005271472.1 | 3 - 4 | 259 | 70 | ||
XM_011542639.2 | XP_011540941.1 | 4 - 5 | 296 | 70 | ||
XM_011542640.2 | XP_011540942.1 | 3 - 4 | 259 | 70 | ||
XM_011542642.1 | XP_011540944.1 | 3 - 4 | 259 | 70 | ||
XM_011542643.2 | XP_011540945.1 | 3 - 4 | 259 | 70 | ||
XM_017017246.1 | XP_016872735.1 | 3 - 4 | 203 | 70 | ||
XM_017017247.1 | XP_016872736.1 | 3 - 4 | 259 | 70 | ||
XM_017017248.1 | XP_016872737.1 | 3 - 4 | 259 | 70 | ||
GenBank mRNA | BC029536.1 | - | 3 - 4 | 216 | 5298158 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002266.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027164.hg.1 | ENST00000393084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027164.hg.1 | ENST00000529391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027164.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027164.hg.1 | TC11002266.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194060.hg.1 | TC1100012227.hg.1 | Clariom D TC |
