Assay Details
| Gene Symbol: LINC00305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027245.1 | 1 - 2 | 252 | 113 | ||
| GenBank mRNA | BC029565.1 | - | 1 - 2 | 256 | 113 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000559.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005840.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005840.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005840.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005840.hg.1 | TC18000559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005844.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005844.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005844.hg.1 | TC18000559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005841.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005841.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005841.hg.1 | TC18000559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005843.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005843.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005843.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005843.hg.1 | TC18000559.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800161299.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161299.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161299.hg.1 | TC1800008920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800161299.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161299.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | TC1800008920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161298.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | TC1800008920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161304.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000454647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | ENST00000456505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008920.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161305.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161305.hg.1 | NR_027245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800161305.hg.1 | TC1800008920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800161305.hg.1 | ENST00000323355.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800161305.hg.1 | NR_027245.1 | Refseq |

LINC00305
Genomic Map