Assay Details
Gene Symbol: ERICH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308234.1 | NP_001295163.1 | 8 - 9 | 808 | 64 | |
NM_152394.4 | NP_689607.2 | 8 - 9 | 1112 | 64 | ||
XM_005247120.3 | XP_005247177.1 | 8 - 9 | 1704 | 64 | ||
GenBank mRNA | AK093375.1 | - | 9 - 10 | 947 | 64 | |
AL834214.1 | - | 612 | 64 | |||
BC029577.1 | - | 8 - 9 | 792 | 5268183 | 64 | |
BC036239.1 | - | 8 - 9 | 1096 | 5297417 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001900.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034243.hg.1 | TC03001900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03034243.hg.1 | ENST00000295910.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034243.hg.1 | ENST00000313811.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034243.hg.1 | ENST00000491361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034243.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300195779.hg.1 | TC0300012804.hg.1 | Clariom D TC |
