Assay Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029605.1 | - | 4 - 5 | 1191 | 5296925 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147284.hg.1 | TC1100006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147285.hg.1 | TC1100006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100147287.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147287.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147287.hg.1 | TC1100006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100147283.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147283.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147283.hg.1 | TC1100006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100147288.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147288.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147288.hg.1 | TC1100006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006756.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000318881.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000524820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | ENST00000532592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | NM_175733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100147286.hg.1 | TC1100006756.hg.1 | Clariom D TC |

SYT9
Genomic Map