Assay Details
| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198475.2 | NP_940877.2 | 5 - 6 | 934 | 75 | |
| XM_017024490.1 | XP_016879979.1 | 3 - 4 | 501 | 75 | ||
| GenBank mRNA | BC029298.1 | - | 5 - 6 | 950 | 75 | |
| BC030200.1 | - | 5 - 6 | 934 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001580.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17021264.hg.1 | ENST00000293443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021264.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021264.hg.1 | TC17001580.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186657.hg.1 | TC1700010828.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186656.hg.1 | TC1700010828.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000293443.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000588067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000589407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | ENST00000592560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010828.hg.1 | NM_198475.2 | Refseq |

FAM171A2
Genomic Map