Assay Details
| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173637.3 | NP_775908.2 | 2 - 3 | 453 | 61 | |
| NR_135612.1 | 2 - 3 | 453 | 61 | |||
| XM_011527926.1 | XP_011526228.1 | 2 - 3 | 247 | 61 | ||
| GenBank mRNA | BC031671.1 | - | 2 - 3 | 453 | 5164630 | 61 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001096.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC031671.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015027.hg.1 | ENST00000321510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015027.hg.1 | ENST00000458275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015027.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015027.hg.1 | TC19001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015030.hg.1 | ENST00000321510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015030.hg.1 | ENST00000458275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015030.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015030.hg.1 | TC19001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015029.hg.1 | ENST00000321510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015029.hg.1 | ENST00000458275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015029.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015029.hg.1 | TC19001096.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | TC1900009430.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175689.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | TC1900009430.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175692.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | TC1900009430.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175693.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | TC1900009430.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900175694.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000321510.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000458275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | ENST00000597558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009430.hg.1 | NM_173637.3 | Refseq |

SLC25A41
Genomic Map