Assay Details
| Gene Symbol: SPINK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC032003.1 | - | 2 - 3 | 335 | 4754193 | 84 |
| BE122852.1 | - | 2 - 3 | 171 | 84 | ||
| GQ504704.1 | - | 2 - 3 | 400 | 84 | ||
| GQ504705.1 | - | 2 - 3 | 336 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162902.hg.1 | TC0500009033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162903.hg.1 | TC0500009033.hg.1 | Clariom D TC |

SPINK6
Genomic Map