Assay Details
| Gene Symbol: PCMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286782.1 | NP_001273711.1 | 2 - 3 | 430 | 143 | |
| NM_001286783.1 | NP_001273712.1 | 4 - 5 | 725 | 143 | ||
| NM_052937.3 | NP_443169.2 | 4 - 5 | 935 | 143 | ||
| XM_005251146.2 | XP_005251203.1 | 4 - 5 | 935 | 143 | ||
| XM_011517444.2 | XP_011515746.1 | 3 - 4 | 524 | 143 | ||
| XM_017013017.1 | XP_016868506.1 | 4 - 5 | 768 | 143 | ||
| GenBank mRNA | AK056952.1 | - | 4 - 5 | 785 | 143 | |
| AK304196.1 | - | 1 - 2 | 208 | 143 | ||
| AK304332.1 | - | 4 - 5 | 509 | 143 | ||
| AL833027.1 | - | 3 - 4 | 336 | 143 | ||
| BC010693.1 | - | 2 - 3 | 218 | 143 | ||
| BC032670.1 | - | 3 - 4 | 680 | 5539318 | 143 | |
| BP418683.1 | - | 3 - 4 | 334 | 143 | ||
| BX640885.1 | - | 2 - 3 | 245 | 143 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001205.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304332.1,NM_001286783.1,XM_011517444.2,XM_017013017.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | TC08001205.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | ENST00000360540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | ENST00000522514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | ENST00000544451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015495.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000519975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010482.hg.1 | ENST00000451824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170697.hg.1 | TC0800010417.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010482.hg.1 | ENST00000451824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170699.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170699.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170699.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170699.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170699.hg.1 | TC0800010417.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000360540.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000519975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000521344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000522514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | ENST00000544451.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_001286783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010417.hg.1 | NM_052937.3 | Refseq |

PCMTD1
Genomic Map